2-((4-Fluorophenyl)amino)propanoic acid 97%
Dodavatel: Apollo Scientific
PC201163-100MG
PC201163-1G
PC201163-250MG
APOSPC201163-100MGEA
827
CZK
APOSPC201163-100MG
APOSPC201163-1G
APOSPC201163-250MG
2-((4-Fluorophenyl)amino)propanoic acid 97%
2-((4-Fluorophenyl)amino)propanoic acid
Vzorec:
C₉H₁₀FNO₂ MW: 183,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04116920 CAS číslo: 16583-82-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...