Tisk…

6-(Trifluoromethyl)quinazolin-4(1H)-one 97%

Dodavatel: Apollo Scientific

PC201162-100MG PC201162-250MG
APOSPC201162-100MGEA 3440 CZK
APOSPC201162-100MG APOSPC201162-250MG
6-(Trifluoromethyl)quinazolin-4(1H)-one 97%
6-(Trifluoromethyl)quinazolin-4(1H)-one
Vzorec: C₉H₅F₃N₂O
Storage Temperature: Ambient
MDL Number: MFCD05664875
CAS číslo: 16544-67-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR