2,2,2-Trifluoro-1-(pyridin-3-yl)ethanone hydrochloride 95+%
Dodavatel: Apollo Scientific
PC201155-100MG
PC201155-1G
PC201155-250MG
APOSPC201155-100MGEA
3380
CZK
APOSPC201155-100MG
APOSPC201155-1G
APOSPC201155-250MG
2,2,2-Trifluoro-1-(pyridin-3-yl)ethanone hydrochloride 95+%
2,2,2-Trifluoro-1-(pyridin-3-yl)ethanone hydrochloride
Vzorec:
C₇H₄F₃NO·ClH MW: 211,569 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD27923425 CAS číslo: 1588441-22-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...