4-Chloro-6-methyl-2-trifluoromethylpyrimidine 97%
Dodavatel: Apollo Scientific
PC201154-100MG
PC201154-1G
PC201154-250MG
APOSPC201154-100MGEA
1910
CZK
APOSPC201154-100MG
APOSPC201154-1G
APOSPC201154-250MG
4-Chloro-6-methyl-2-trifluoromethylpyrimidine 97%
4-Chloro-6-methyl-2-trifluoromethylpyrimidine
Vzorec:
C₆H₄ClF₃N₂ Storage Temperature: Ambient |
MDL Number:
MFCD09743686 CAS číslo: 1582-25-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...