Tisk…

(S)-Methyl 4,4-difluoropyrrolidine-2-carboxylate hydrochloride 95+%

Dodavatel: Apollo Scientific

Synonyms: N-4,4-DiF-Pro-OMe·HCl

PC201152-1G PC201152-250MG
APOSPC201152-1GEA 8800 CZK
APOSPC201152-1G APOSPC201152-250MG
(S)-Methyl 4,4-difluoropyrrolidine-2-carboxylate hydrochloride 95+%
(S)-Methyl 4,4-difluoropyrrolidine-2-carboxylate hydrochloride
Vzorec: C₆H₉F₂NO₂·ClH
MW: 201,599 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23106100
CAS číslo: 156046-05-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR