Tisk…

2-Chloro-3,4-difluorobenzoic acid 95+%

Dodavatel: Apollo Scientific

PC201146-1G PC201146-250MG
APOSPC201146-1GEA 5630 CZK
APOSPC201146-1G APOSPC201146-250MG
2-Chloro-3,4-difluorobenzoic acid 95+%
2-Chloro-3,4-difluorobenzoic acid
Vzorec: C₇H₃ClF₂O₂
MW: 192,547 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18415556
CAS číslo: 150444-93-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR