4-Bromo-2-fluoro-6-methoxybenzoic acid 97%
Dodavatel: Apollo Scientific
PC201145-100MG
PC201145-250MG
PC201145-1G
APOSPC201145-100MGEA
0
CZK
APOSPC201145-100MG
APOSPC201145-250MG
APOSPC201145-1G
4-Bromo-2-fluoro-6-methoxybenzoic acid 97%
4-Bromo-2-fluoro-6-methoxybenzoic acid
Vzorec:
C₈H₆BrFO₃ Storage Temperature: Ambient |
MDL Number:
MFCD22570393 CAS číslo: 1472104-49-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...