Tisk…

3-Bromo-6-fluoro-2-methylbenzoic acid 95+%

Dodavatel: Apollo Scientific

PC201139-100MG PC201139-250MG PC201139-1G
APOSPC201139-100MGEA 0 CZK
APOSPC201139-100MG APOSPC201139-250MG APOSPC201139-1G
3-Bromo-6-fluoro-2-methylbenzoic acid 95+%
3-Bromo-6-fluoro-2-methylbenzoic acid
Vzorec: C₈H₆BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD23712078
CAS číslo: 1427373-55-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR