Tisk…

5-Bromo-3-fluoro-1-methylpyridin-2(1H)-one 98%

Dodavatel: Apollo Scientific

PC201127-100MG PC201127-250MG
APOSPC201127-100MGEA 827 CZK
APOSPC201127-100MG APOSPC201127-250MG
5-Bromo-3-fluoro-1-methylpyridin-2(1H)-one 98%
5-Bromo-3-fluoro-1-methylpyridin-2(1H)-one
Vzorec: C₆H₅BrFNO
MW: 206.012 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23135714
CAS číslo: 1352152-46-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR