Tisk…

Methyl 5-fluoro-3-methylpicolinate 98%

Dodavatel: Apollo Scientific

PC201123-100MG PC201123-1G PC201123-250MG
APOSPC201123-100MGEA 2040 CZK
APOSPC201123-100MG APOSPC201123-1G APOSPC201123-250MG
Methyl 5-fluoro-3-methylpicolinate 98%
Methyl 5-fluoro-3-methylpicolinate
Vzorec: C₈H₈FNO₂
Storage Temperature: Ambient
MDL Number: MFCD22560016
CAS číslo: 1346148-32-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR