(R)-6-Fluorochroman-2-carboxylic acid 95+%
Dodavatel: Apollo Scientific
PC201114-1G
PC201114-5G
APOSPC201114-1GEA
870
CZK
APOSPC201114-1G
APOSPC201114-5G
(R)-6-Fluorochroman-2-carboxylic acid 95+%
Vzorec:
C₁₀H₉FO₃ Storage Temperature: Ambient |
MDL Number:
MFCD09955074 CAS číslo: 129101-37-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...