Tisk…

(4-Fluorotetrahydro-2H-pyran-4-yl)methanamine hydrochloride 97%

Dodavatel: Apollo Scientific

PC201104-100MG PC201104-1G PC201104-250MG
APOSPC201104-100MGEA 3740 CZK
APOSPC201104-100MG APOSPC201104-1G APOSPC201104-250MG
(4-Fluorotetrahydro-2H-pyran-4-yl)methanamine hydrochloride 97%
(4-Fluorotetrahydro-2H-pyran-4-yl)methanamine hydrochloride
Vzorec: C₆H₁₂FNO·ClH
MW: 169,625 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18791648
CAS číslo: 1263180-53-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR