Tisk…

2-Fluoro-3-(hydroxymethyl)benzonitrile 95+%

Dodavatel: Apollo Scientific
Varování

PC201103-100MG PC201103-250MG
APOSPC201103-100MGEA 1120 CZK
APOSPC201103-100MG APOSPC201103-250MG
2-Fluoro-3-(hydroxymethyl)benzonitrile 95+%
2-Fluoro-3-(hydroxymethyl)benzonitrile
Vzorec: C₈H₆FNO
MW: 151,138 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18394036
CAS číslo: 1261861-91-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR