5-Amino-2,3-difluorobenzonitrile 97%
Dodavatel: Apollo Scientific
PC201092-1G
PC201092-250MG
PC201092-5G
APOSPC201092-1GEA
7840
CZK
APOSPC201092-1G
APOSPC201092-250MG
APOSPC201092-5G
5-Amino-2,3-difluorobenzonitrile 97%
5-Amino-2,3-difluorobenzonitrile
Vzorec:
C₇H₄F₂N₂ MW: 154,117 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD20726994 CAS číslo: 1247885-41-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...