Tisk…

3-Bromo-2-(difluoromethoxy)pyridine 97%

Dodavatel: Apollo Scientific

PC201073-100MG PC201073-1G PC201073-250MG
APOSPC201073-100MGEA 3940 CZK
APOSPC201073-100MG APOSPC201073-1G APOSPC201073-250MG
3-Bromo-2-(difluoromethoxy)pyridine 97%
3-Bromo-2-(difluoromethoxy)pyridine
Vzorec: C₆H₄BrF₂NO
MW: 224,003 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185844
CAS číslo: 1214345-30-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR