Tisk…

(S)-1-(4-Fluoro-3-methylphenyl)ethanamine 95+%

Dodavatel: Apollo Scientific

PC201067-100MG PC201067-1G PC201067-250MG
APOSPC201067-100MGEA 3380 CZK
APOSPC201067-100MG APOSPC201067-1G APOSPC201067-250MG
(S)-1-(4-Fluoro-3-methylphenyl)ethanamine 95+%
(S)-1-(4-Fluoro-3-methylphenyl)ethanamine
Vzorec: C₉H₁₂FN·ClH
Storage Temperature: Ambient
MDL Number: MFCD08057493
CAS číslo: 1213299-63-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR