Tisk…

4-Bromo-5-fluoro-2-methylpyridine 97%

Dodavatel: Apollo Scientific

PC201066-1G PC201066-250MG
APOSPC201066-1GEA 10220 CZK
APOSPC201066-1G APOSPC201066-250MG
4-Bromo-5-fluoro-2-methylpyridine 97%
4-Bromo-5-fluoro-2-methylpyridine
Vzorec: C₆H₅BrFN
MW: 190.013 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18251020
CAS číslo: 1211590-24-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR