6-(Difluoromethoxy)nicotinaldehyde 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC201062-100MG
PC201062-1G
PC201062-250MG
APOSPC201062-100MGEA
4100
CZK
APOSPC201062-100MG
APOSPC201062-1G
APOSPC201062-250MG
6-(Difluoromethoxy)nicotinaldehyde 98%
6-(Difluoromethoxy)nicotinaldehyde
Vzorec:
C₇H₅F₂NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD18261899 CAS číslo: 1211577-99-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...