Tisk…

2-Bromo-4-(difluoromethoxy)pyridine 95+%

Dodavatel: Apollo Scientific

PC201054-100MG PC201054-250MG
APOSPC201054-100MGEA 5890 CZK
APOSPC201054-100MG APOSPC201054-250MG
2-Bromo-4-(difluoromethoxy)pyridine 95+%
2-Bromo-4-(difluoromethoxy)pyridine
Vzorec: C₆H₄BrF₂NO
MW: 224,003 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185841
CAS číslo: 1206984-48-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR