Tisk…
4,6-Difluoro-2,3-dihydro-1H-inden-1-amine hydrochloride 97%
Dodavatel: Apollo Scientific
PC201049-100MG PC201049-1G PC201049-250MG
APOSPC201049-100MGEA 2860 CZK
APOSPC201049-100MG APOSPC201049-1G APOSPC201049-250MG
4,6-Difluoro-2,3-dihydro-1H-inden-1-amine hydrochloride 97%
4,6-Difluoro-2,3-dihydro-1H-inden-1-amine hydrochloride
Vzorec: C₉H₉F₂N·ClH
MW: 205,632 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08544172
CAS číslo: 1199782-88-1