3-Bromo-5-fluoro-4-methylbenzoic acid 95+%
Dodavatel: Apollo Scientific
PC201046-10G
PC201046-1G
PC201046-5G
APOSPC201046-10GEA
23310
CZK
APOSPC201046-10G
APOSPC201046-1G
APOSPC201046-5G
3-Bromo-5-fluoro-4-methylbenzoic acid 95+%
3-Bromo-5-fluoro-4-methylbenzoic acid
Vzorec:
C₈H₆BrFO₂ MW: 233,034 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19443258 CAS číslo: 1191988-29-0 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...