5-Bromo-3-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine 95+%
Dodavatel: Apollo Scientific
PC201031-100MG
PC201031-1G
PC201031-250MG
APOSPC201031-100MGEA
3740
CZK
APOSPC201031-100MG
APOSPC201031-1G
APOSPC201031-250MG
5-Bromo-3-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine 95+%
5-Bromo-3-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine
Vzorec:
C₈H₄BrF₃N₂ MW: 265,03 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12024503 CAS číslo: 1150618-36-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...