Tisk…

2-Chloro-4-fluoronicotinonitrile 95%

Dodavatel: Apollo Scientific
Varování

PC201012-1G PC201012-250MG
APOSPC201012-1GEA 17960 CZK
APOSPC201012-1G APOSPC201012-250MG
2-Chloro-4-fluoronicotinonitrile 95%
2-Chloro-4-fluoronicotinonitrile
Vzorec: C₆H₂ClFN₂
MW: 156,545 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27996745
CAS číslo: 1054552-27-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR