4-Amino-6-fluoropyridin-2(1H)-one 95+%
Dodavatel: Apollo Scientific
PC201010-100MG
PC201010-250MG
APOSPC201010-100MGEA
870
CZK
APOSPC201010-100MG
APOSPC201010-250MG
4-Amino-6-fluoropyridin-2(1H)-one 95+%
4-Amino-6-fluoropyridin-2(1H)-one
Vzorec:
C₅H₅FN₂O MW: 128,104 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18427750 CAS číslo: 105252-99-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...