Tisk…
(4-(Trifluoromethyl)pyridin-2-yl)methanamine hydrochloride 96%
Dodavatel: Apollo Scientific
PC201004-1G PC201004-250MG PC201004-5G
APOSPC201004-1GEA 7640 CZK
APOSPC201004-1G APOSPC201004-250MG APOSPC201004-5G
(4-(Trifluoromethyl)pyridin-2-yl)methanamine hydrochloride 96%
(4-(Trifluoromethyl)pyridin-2-yl)methanamine hydrochloride
Vzorec: C₇H₇F₃N₂·ClH
MW: 212.6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04117763
CAS číslo: 1005515-26-3