Tisk…

5-Fluoro-6-methylpyridine-2-carboxylic acid 97%

Dodavatel: Apollo Scientific

PC201003-1G PC201003-250MG
APOSPC201003-1GEA 5260 CZK
APOSPC201003-1G APOSPC201003-250MG
5-Fluoro-6-methylpyridine-2-carboxylic acid 97%
5-Fluoro-6-methylpyridine-2-carboxylic acid
Vzorec: C₇H₆FNO₂
MW: 155.126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11040267
CAS číslo: 1005474-88-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR