Tisk…

2-Bromo-4-fluoro-5-methylbenzoic acid 97%

Dodavatel: Apollo Scientific

PC201001-10G PC201001-1G PC201001-5G
APOSPC201001-10GEA 23050 CZK
APOSPC201001-10G APOSPC201001-1G APOSPC201001-5G
2-Bromo-4-fluoro-5-methylbenzoic acid 97%
2-Bromo-4-fluoro-5-methylbenzoic acid
Vzorec: C₈H₆BrFO₂
MW: 233,034 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09263434
CAS číslo: 1003709-39-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR