(S)-1-(2-Chloro-6-fluorophenyl)ethanamine hydrochloride 96%
Dodavatel: Apollo Scientific
PC201000-1G
PC201000-250MG
APOSPC201000-1GEA
21760
CZK
APOSPC201000-1G
APOSPC201000-250MG
(S)-1-(2-Chloro-6-fluorophenyl)ethanamine hydrochloride 96%
(S)-1-(2-Chloro-6-fluorophenyl)ethanamine hydrochloride
Vzorec:
C₈H₉ClFN·ClH Storage Temperature: Ambient |
MDL Number:
MFCD12757994 CAS číslo: 1000878-48-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...