Tisk…

Ethyl 4-(trifluoromethyl)pyrimidine-5-carboxylate

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-(Ethoxycarbonyl)-4-(trifluoromethyl)-1,3-diazine, 5-(Ethoxycarbonyl)-4-(trifluoromethyl)pyrimidine

PC200538-1G PC200538-5G
APOSPC200538-1GEA 4140 CZK
APOSPC200538-1G APOSPC200538-5G
Ethyl 4-(trifluoromethyl)pyrimidine-5-carboxylate
Ethyl 4-(trifluoromethyl)pyrimidine-5-carboxylate
Vzorec: C₈H₇F₃N₂O₂
MW: 220,149 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07367356
CAS číslo: 187035-81-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR