Tisk…

(Z)-N'-Hydroxy-2-(trifluoromethyl)benzene-1-carboximidamide 97%

Dodavatel: Apollo Scientific

PC200395-5G PC200395-1G
APOSPC200395-5GEA 2690 CZK
APOSPC200395-5G APOSPC200395-1G
(Z)-N'-Hydroxy-2-(trifluoromethyl)benzene-1-carboximidamide 97%
(Z)-N'-Hydroxy-2-(trifluoromethyl)benzene-1-carboximidamide
Vzorec: C₈H₇F₃N₂O
MW: 204,149 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660165
CAS číslo: 40067-66-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR