Tisk…

2-Chloro-4-(4-fluoro-2-methoxyphenyl)pyridine

Dodavatel: Apollo Scientific

PC200390-1G PC200390-500MG
APOSPC200390-1GEA 19110 CZK
APOSPC200390-1G APOSPC200390-500MG
2-Chloro-4-(4-fluoro-2-methoxyphenyl)pyridine
2-Chloro-4-(4-fluoro-2-methoxyphenyl)pyridine
Vzorec: C₁₂H₉ClFNO
MW: 237.657 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26386176
CAS číslo: 1333468-54-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR