Tisk…

Potassium 6-chloropyridine-3-trifluoroborate

Dodavatel: Apollo Scientific

PC200191-1G PC200191-500MG PC200191-5G
APOSPC200191-1GEA 4770 CZK
APOSPC200191-1G APOSPC200191-500MG APOSPC200191-5G
Potassium 6-chloropyridine-3-trifluoroborate
Potassium 6-chloropyridine-3-trifluoroborate
Vzorec: C₅H₃BClF₃N·K
Storage Temperature: Ambient
MDL Number: MFCD09993185
CAS číslo: 1235099-38-3

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR