Tisk…

Potassium (3-(hydroxymethyl) phenyl)trifluoroborate

Dodavatel: Apollo Scientific

PC200176-1G PC200176-500MG PC200176-5G
APOSPC200176-1GEA 3340 CZK
APOSPC200176-1G APOSPC200176-500MG APOSPC200176-5G
Potassium (3-(hydroxymethyl) phenyl)trifluoroborate
Potassium (3-(hydroxymethyl) phenyl)trifluoroborate
Vzorec: C₇H₇BF₃O·K
MW: 214,034 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09993091
CAS číslo: 1015082-77-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR