3-(Difluoromethyl)-2-fluoro-4-iodopyridine
Dodavatel: Apollo Scientific
PC200122-1G
PC200122-500MG
APOSPC200122-1GEA
6980
CZK
APOSPC200122-1G
APOSPC200122-500MG
3-(Difluoromethyl)-2-fluoro-4-iodopyridine
3-(Difluoromethyl)-2-fluoro-4-iodopyridine
Vzorec:
C₆H₃F₃IN Storage Temperature: Ambient |
MDL Number:
MFCD25479865 CAS číslo: 1446786-38-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...