Potassium trifluoro(hex-1-yn-1-yl)borate
Dodavatel: Apollo Scientific
PC200095-1G
PC200095-500MG
APOSPC200095-1GEA
5330
CZK
APOSPC200095-1G
APOSPC200095-500MG
Potassium trifluoro(hex-1-yn-1-yl)borate
Potassium trifluoro(hex-1-yn-1-yl)borate
Vzorec:
C₆H₉BF₃·K MW: 188,04 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11052661 CAS číslo: 244301-59-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...