4,6-Dichloro-5-(difluoromethyl)pyrimidine 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC200087-1G
PC200087-500MG
APOSPC200087-1GEA
8100
CZK
APOSPC200087-1G
APOSPC200087-500MG
4,6-Dichloro-5-(difluoromethyl)pyrimidine 97%
4,6-Dichloro-5-(difluoromethyl)pyrimidine
Vzorec:
C₅H₂Cl₂F₂N₂ MW: 198,986 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD25368744 CAS číslo: 1443290-45-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...