2,4-Dichloro-3-(difluoromethyl)pyridine
Dodavatel: Apollo Scientific
PC200085-1G
PC200085-500MG
APOSPC200085-1GEA
6880
CZK
APOSPC200085-1G
APOSPC200085-500MG
2,4-Dichloro-3-(difluoromethyl)pyridine
2,4-Dichloro-3-(difluoromethyl)pyridine
Vzorec:
C₆H₃Cl₂F₂N Storage Temperature: Ambient |
MDL Number:
MFCD25368742 CAS číslo: 1443290-44-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...