2-Chloro-4-[(2,2,2-trifluoroethyl)amino]pyrimidine-5-carbonitrile
Dodavatel: Apollo Scientific
PC200057-1G
PC200057-500MG
APOSPC200057-1GEA
15310
CZK
APOSPC200057-1G
APOSPC200057-500MG
2-Chloro-4-[(2,2,2-trifluoroethyl)amino]pyrimidine-5-carbonitrile
2-Chloro-4-[(2,2,2-trifluoroethyl)amino]pyrimidine-5-carbonitrile
Vzorec:
C₇H₄ClF₃N₄ MW: 236,582 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD23697507 CAS číslo: 389606-51-3 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...