2-Chloro-5-fluoro-4-(methylsulphanyl)pyrimidine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Chloro-5-fluoro-4-(methylthio)pyrimidine
PC200050-1G
PC200050-250MG
APOSPC200050-1GEA
744
CZK
APOSPC200050-1G
APOSPC200050-250MG
2-Chloro-5-fluoro-4-(methylsulphanyl)pyrimidine
2-Chloro-5-fluoro-4-(methylsulphanyl)pyrimidine
Vzorec:
C₅H₄ClFN₂S MW: 178.615 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16876993 CAS číslo: 87789-51-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...