Tisk…

5-Bromo-3-chloro-6-(trifluoromethyl)-1H-indole

Dodavatel: Apollo Scientific

PC200027-1G PC200027-500MG
APOSPC200027-1GEA 15010 CZK
APOSPC200027-1G APOSPC200027-500MG
5-Bromo-3-chloro-6-(trifluoromethyl)-1H-indole
5-Bromo-3-chloro-6-(trifluoromethyl)-1H-indole
Vzorec: C₉H₄BrClF₃N
Bod tání: 88…91 °C
Storage Temperature: Ambient
MDL Number: MFCD22689147
CAS číslo: 1420537-62-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR