Tisk…

3-Amino-5-chloro-4-hydroxybenzotrifluoride 98%

Dodavatel: Apollo Scientific
Varování

PC1990-250MG PC1990-1G
APOSPC1990-250MGEA 4960 CZK
APOSPC1990-250MG APOSPC1990-1G
3-Amino-5-chloro-4-hydroxybenzotrifluoride 98%
3-Amino-5-chloro-4-hydroxybenzotrifluoride
Vzorec: C₇H₅ClF₃NO
Storage Temperature: Ambient
MDL Number: MFCD07777110
CAS číslo: 78068-81-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR