Aluminium trifluoroacetate 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC1976-1G
PC1976-5G
APOSPC1976-1GEA
3340
CZK
APOSPC1976-1G
APOSPC1976-5G
Aluminium trifluoroacetate 97%
Aluminium trifluoroacetate
Vzorec:
Al·3CF₃CO₂ MW: 366,028 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03411227 CAS číslo: 36554-89-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...