Ethyl 3-oxo-3-[4-(trifluoromethyl)phenyl]propanoate
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Ethyl [4-(trifluoromethyl)benzoyl]acetate, Ethyl 3-oxo-3-(4-(trifluoromethyl)phenyl)propanoate
PC1854-5G
APOSPC1854-5GEA
2690
CZK
APOSPC1854-5G
Ethyl 3-oxo-3-[4-(trifluoromethyl)phenyl]propanoate
Ethyl 3-oxo-3-[4-(trifluoromethyl)phenyl]propanoate
Vzorec:
C₁₂H₁₁F₃O₃ MW: 260,209 g/mol Bod varu: 248…249 °C Hustota: 1,27 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD03424818 CAS číslo: 106263-53-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...