4-(Difluoromethyl)benzeneboronic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(4-(Difluoromethyl)phenyl)boronic acid, 4-Boronobenzal fluoride
PC1598-1G
PC1598-250MG
PC1598-5G
APOSPC1598-1GEA
2030
CZK
APOSPC1598-1G
APOSPC1598-250MG
APOSPC1598-5G
4-(Difluoromethyl)benzeneboronic acid
4-(Difluoromethyl)benzeneboronic acid
Vzorec:
C₇H₇BF₂O₂ MW: 171,937 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09056705 CAS číslo: 946525-43-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...