2-Fluorophenylacetyl chloride
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC1581-25G
PC1581-5G
APOSPC1581-25GEA
7810
CZK
APOSPC1581-25G
APOSPC1581-5G
2-Fluorophenylacetyl chloride
2-Fluorophenylacetyl chloride
Vzorec:
C₈H₆ClFO Bod varu: 203…204 °C Storage Temperature: Ambient |
MDL Number:
MFCD03840709 CAS číslo: 451-81-0 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...