Tisk…

Perfluoro-2-bromopropanoyl bromide 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC1567N-10G PC1567N-5G
APOSPC1567N-10GEA 9960 CZK
APOSPC1567N-10G APOSPC1567N-5G
Perfluoro-2-bromopropanoyl bromide 97%
Perfluoro-2-bromopropanoyl bromide
Vzorec: C₃Br₂F₄O
Bod varu: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00153722
CAS číslo: 41874-81-1
UN: 1760
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR