Tisk…

Methyl 2,3,4,5-tetrafluorobenzoate

Dodavatel: Apollo Scientific
Varování

Synonyms: Methyl 2H-tetrafluorobenzoate

PC1521-1G PC1521-25G PC1521-5G
APOSPC1521-1GEA 662 CZK
APOSPC1521-1G APOSPC1521-25G APOSPC1521-5G
Methyl 2,3,4,5-tetrafluorobenzoate
Methyl 2,3,4,5-tetrafluorobenzoate
Vzorec: C₈H₄F₄O₂
MW: 208,11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06203829
CAS číslo: 5292-42-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR