Tisk…

5-Chloro-2,4,6-trifluoroisophthalonitrile

Dodavatel: Apollo Scientific
Varování

PC1506-1G PC1506-250MG PC1506-5G
APOSPC1506-1GEA 13530 CZK
APOSPC1506-1G APOSPC1506-250MG APOSPC1506-5G
5-Chloro-2,4,6-trifluoroisophthalonitrile
5-Chloro-2,4,6-trifluoroisophthalonitrile
Vzorec: C₈ClF₃N₂
MW: 216,547 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06658269
CAS číslo: 1897-50-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR