Tisk…

1-Bromo-2-methyl-4,4,4-trifluorobutane

Dodavatel: Apollo Scientific
Varování

PC1487-1G PC1487-250MG
APOSPC1487-1GEA 9330 CZK
APOSPC1487-1G APOSPC1487-250MG
1-Bromo-2-methyl-4,4,4-trifluorobutane
1-Bromo-2-methyl-4,4,4-trifluorobutane
Vzorec: C₅H₈BrF₃
MW: 205,016 g/mol
Bod varu: 115…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00190632
CAS číslo: 203302-90-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR