4-Chloro-2-fluoro-5-sulphamoylbenzoic acid
Dodavatel: Apollo Scientific
PC1482-10G
PC1482-1G
PC1482-5G
APOSPC1482-10GEA
6020
CZK
APOSPC1482-10G
APOSPC1482-1G
APOSPC1482-5G
4-Chloro-2-fluoro-5-sulphamoylbenzoic acid
4-Chloro-2-fluoro-5-sulphamoylbenzoic acid
Vzorec:
C₇H₅ClFNO₄S Storage Temperature: Ambient |
MDL Number:
MFCD03265327 CAS číslo: 4793-22-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...